Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002824.3 | 26 | 4034 | |||
GenBank mRNA | AF041080.1 | - | LIT2190 HERC2P2 |
26 | 4040 | |
BC128547.1 | - | 3 | 308 | |||
CR749409.1 | - | 15 | 2476 |