Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026955.1 | 3 | 1980 | |||
GenBank mRNA | AK001938.1 | - | 2 | 3150 | ||
AK001941.1 | - | 3 | 1827 |