Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002719.2 | 1 | 1203 | |||
GenBank mRNA | M55632.1 | - | 1 | 1202 |
Gene Symbol: FMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509347.2 | XP_011507649.1 | 10 | 2556 |