Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258026.1 | NP_001244955.1 | 1 | 268 | ||
GenBank mRNA | AK298809.1 | - | 1 | 269 |