Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003064.2 | 2 | 668 | |||
GenBank mRNA | AF222904.1 | - | 2 | 417 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX116346.1 | - | 1 | 45 | Chr. 17 - 28227613 - 28228047 [+] on Build GRCh38 |