Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTTY15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001545.2 | 4 | 1403 |
Gene Symbol: MICAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028867.1 | XP_016884356.1 | 1 | 513 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL080135.3 | - | 4 | 1404 | Chr. Y - 12662367 - 12692230 [+] on Build GRCh38 |