Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC154761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015421.1 | 3 | 1679 | |||
GenBank mRNA | AK124492.1 | - | 1 | 2442 | ||
BC041464.1 | - | 2 | 1042 |
Gene Symbol: LOC101928422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012897.1 | XP_016868386.1 | 5 | 2112 |