Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGLUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024265.1 | 5 | 625 | |||
XM_017006878.1 | XP_016862367.1 | 5 | 597 | |||
XM_017006879.1 | XP_016862368.1 | 6 | 743 | |||
GenBank mRNA | AK295984.1 | - | 5 | 600 | ||
AK301002.1 | - | 6 | 741 | |||
CD517010.1 | - | 5 | 541 |