Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URAHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027335.2 | 4 | 422 | |||
NR_027336.2 | 4 | 375 | ||||
GenBank mRNA | AK304171.1 | - | 4 | 365 | ||
BC044232.1 | - | 4 | 422 | |||
BC089450.1 | - | 4 | 358 | |||
DC393803.1 | - | 4 | 376 |
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC036781.1 | - | 1 | 957 |