Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027779.1 | 4, 5 | 428 | |||
XM_011530108.2 | XP_011528410.1 | 4, 5 | 383 | |||
GenBank mRNA | AF173935.1 | - | 4, 5 | 338 | ||
AL096883.1 | - | 3, 4 | 276 |