Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC441601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003034.2 | 2 | 331 | |||
GenBank mRNA | BC067889.1 | - | 2 | 321 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI341338.1 | - | 1 | 61 | Chr. 11 - 50280052 - 50295338 [-] on Build GRCh38 |