Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027118.1 | 2 | 222 | |||
NR_027119.1 | 2 | 190 | ||||
GenBank mRNA | AK096266.1 | - | 1 | 204 | ||
BC037931.1 | - | 2 | 227 | |||
BC047446.1 | - | 2 | 189 |