Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC728323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130699.1 | 7 | 858 | |||
NR_130700.1 | 5 | 630 | ||||
NR_130701.1 | 7 | 783 | ||||
NR_130702.1 | 6 | 734 | ||||
GenBank mRNA | AK125248.1 | - | 6 | 671 | ||
AK298283.1 | - | 7 | 798 | |||
AK307199.1 | - | 7 | 723 | |||
AK309231.1 | - | 5 | 579 |