Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTAGE10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003268.3 | 1 | 1507 | |||
GenBank mRNA | BC030655.1 | - | 1 | 1406 |
Gene Symbol: CTAGE3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF338231.1 | - | 1 | 776 |