Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000231.1 | XP_016855720.1 | 3 | 2386 | ||
XM_017000232.1 | XP_016855721.1 | 2 | 2192 | |||
GenBank mRNA | AF370397.1 | - | 1 | 1492 | ||
AK055266.1 | - | 3 | 2365 | |||
AK093736.1 | - | 2 | 2195 |