Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323930.1 | NP_001310859.1 | 1 | 96 | ||
NM_001323932.1 | NP_001310861.1 | 1 | 96 | |||
NR_136654.1 | 1 | 96 | ||||
XM_017000231.1 | XP_016855720.1 | 1 | 96 | |||
XM_017000232.1 | XP_016855721.1 | 1 | 65 | |||
GenBank mRNA | AK055266.1 | - | 1 | 75 | ||
AK093736.1 | - | 1 | 68 |