Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028137.1 | 2 | 434 | |||
GenBank mRNA | BC022493.1 | - | 2 | 385 | ||
BC037315.1 | - | 2 | 353 | |||
BC047092.1 | - | 2 | 358 | |||
BC052342.1 | - | 1 | 1738 | |||
DA291647.1 | - | 2 | 411 | |||
DA295451.1 | - | 2 | 434 |