Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028137.1 | 2 | 1009 | |||
GenBank mRNA | AK091347.1 | - | 1 | 424 | ||
BC022493.1 | - | 2 | 960 | |||
BC037315.1 | - | 2 | 928 | |||
BC047092.1 | - | 2 | 933 |