Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPI1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002187.3 | 1 | 1756 | |||
GenBank mRNA | AK094894.1 | - | 1 | 1758 | ||
BC053624.1 | - | 1 | 1015 |