Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027299.1 | 3, 4 | 516 | |||
GenBank mRNA | AK311039.1 | - | 3, 4 | 445 | ||
KJ535085.1 | - | 3, 4 | 412 |