Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324128.1 | NP_001311057.1 | 1, 2 | 153 | ||
NM_001324132.1 | NP_001311061.1 | 1, 2 | 153 | |||
GenBank mRNA | AK093693.1 | - | 1, 2 | 111 | ||
DA484161.1 | - | 1, 2 | 149 |