Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717528.3 | XP_006717591.3 | 1 | 51 | ||
XM_017016884.1 | XP_016872373.1 | 1 | 51 | |||
XM_017016885.1 | XP_016872374.1 | 1 | 51 | |||
XM_017016886.1 | XP_016872375.1 | 1 | 51 | |||
XM_017016890.1 | XP_016872379.1 | 1 | 51 | |||
XM_017016893.1 | XP_016872382.1 | 1 | 51 | |||
XM_017016894.1 | XP_016872383.1 | 1 | 51 |
Gene Symbol: LRRC37A6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003525.2 | 1 | 3683 | |||
GenBank mRNA | AK126872.1 | - | 1 | 1939 |