Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026863.1 | 2 | 197 | |||
GenBank mRNA | AF426261.1 | - | 2 | 197 | ||
BC107109.2 | - | 2 | 128 |
Gene Symbol: LOC102724354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136540.1 | 3 | 158 | |||
GenBank mRNA | HY162931.1 | - | 2 | 116 |