Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026542.1 | 1 | 105 | |||
NR_026543.1 | 1 | 105 | ||||
GenBank mRNA | BC062296.1 | - | 1 | 107 | ||
BC080530.1 | - | 1 | 104 | |||
BG751912.1 | - | 1 | 105 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU174860.1 | - | 1 | 106 | Chr. 21 - 39606118 - 39612823 [-] on Build GRCh38 |