Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANTXRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278688.2 | NP_001265617.1 | 10, 11 | 1158 | ||
XM_006717684.3 | XP_006717747.1 | 10, 11 | 1158 | |||
XM_011539430.2 | XP_011537732.1 | 10, 11 | 1158 | |||
XM_011539431.2 | XP_011537733.1 | 9, 10 | 840 | |||
XM_011539433.2 | XP_011537735.1 | 9, 10 | 947 | |||
XM_011539434.2 | XP_011537736.1 | 9, 10 | 747 | |||
XM_011539437.2 | XP_011537739.1 | 9, 10 | 1266 | |||
XM_011539438.2 | XP_011537740.1 | 9, 10 | 1262 | |||
XM_011539439.2 | XP_011537741.1 | 9, 10 | 1529 | |||
XM_011539440.2 | XP_011537742.1 | 10, 11 | 1326 | |||
XM_011539441.1 | XP_011537743.1 | 10, 11 | 1158 | |||
XM_011539442.1 | XP_011537744.1 | 10, 11 | 1158 | |||
XM_017015818.1 | XP_016871307.1 | 9, 10 | 752 | |||
XM_017015819.1 | XP_016871308.1 | 9, 10 | 1417 | |||
GenBank mRNA | AK057316.1 | - | 11, 12 | 859 |