Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD19P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026868.1 | 8 | 1000 | |||
GenBank mRNA | AK093497.1 | - | 6 | 592 | ||
AK292218.1 | - | 8 | 1413 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY995235.1 | - | 7 | 685 | Chr. 9 - 92809769 - 92829056 [+] on Build GRCh38 |