Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD19P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026868.1 | 5 | 687 | |||
GenBank mRNA | AK093497.1 | - | 3 | 279 | ||
AK292218.1 | - | 5 | 1100 | |||
BC028712.1 | - | 6 | 877 | |||
BC038951.1 | - | 5 | 687 | 5733799 | ||
BC117367.1 | - | 5 | 662 | 40125918 | ||
BC171729.1 | - | 5 | 662 | 9052104 |