Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM183CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024473.1 | 3 | 2809 | |||
GenBank mRNA | AL833525.1 | - | 3 | 2503 | ||
BC028745.1 | - | 4 | 2340 | |||
BC045833.1 | - | 4 | 2335 |