Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC146880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026899.1 | 3 | 908 | |||
NR_027487.2 | 2 | 696 | ||||
GenBank mRNA | AK055342.1 | - | 2 | 779 | ||
BC032382.1 | - | 2 | 696 | |||
BC120976.1 | - | 2 | 860 | |||
BC120977.2 | - | 2 | 860 | |||
DB093141.1 | - | 3, 4 | 392 |