Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC400927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002821.2 | 2, 3 | 211 | |||
GenBank mRNA | AK124064.1 | - | 2, 3 | 203 | ||
AK307225.1 | - | 2, 3 | 158 | |||
BI669165.1 | - | 2, 3 | 217 | |||
CN357790.1 | - | 2, 3 | 123 |
Gene Symbol: LOC400927-CSNK1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289912.1 | NP_001276841.1 | 2, 3 | 149 | ||
GenBank mRNA | AK092269.1 | - | 2, 3 | 149 | ||
AK310303.1 | - | 2, 3 | 158 |