Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIRLET7BHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027033.2 | 6 | 1917 | |||
NR_110479.1 | 5 | 1766 | ||||
GenBank mRNA | AK023424.1 | - | 6 | 1919 | ||
AK130875.1 | - | 5 | 1760 |