Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF204P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002722.2 | 4 | 1953 | |||
NR_024553.1 | 5 | 833 | ||||
GenBank mRNA | AF033199.1 | - | LIT2063 ZNF204 pseudogene |
3 | 556 | |
AL833520.1 | - | 4 | 1953 | |||
BC067282.1 | - | 2 | 805 |