Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024398.1 | 1 | 41 | |||
GenBank mRNA | AK311179.1 | - | 1 | 72 | ||
CX786597.1 | - | 1 | 98 | |||
DA293448.1 | - | 1 | 42 |