Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028344.1 | 1, 2 | 49 | |||
NR_028345.2 | 1, 2 | 49 | ||||
NR_104228.1 | 1, 2 | 49 | ||||
GenBank mRNA | AK054978.1 | - | 1, 2 | 41 | ||
BM811380.1 | - | 1, 2 | 28 | |||
BX508035.1 | - | 1, 2 | 42 | |||
DA082671.1 | - | 1, 2 | 49 | |||
DA128399.1 | - | 1, 2 | 49 |