Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT14P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027094.1 | 1 | 1220 | |||
GenBank mRNA | AY954513.2 | - | 1 | 1220 | ||
BC132697.1 | - | 1 | 1192 |