Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF217-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026876.1 | 7 | 2852 | |||
GenBank mRNA | AL713649.2 | - | 7 | 2854 |