Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002314.3 | 2 | 861 | |||
GenBank mRNA | AK097753.1 | - | 1 | 1133 | ||
BC128116.1 | - | 2 | 861 | |||
BC128117.1 | - | 2 | 861 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX782089.1 | - | 1 | 310 | Chr. 1 - 53437904 - 53438743 [-] on Build GRCh38 |