Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1DP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027062.1 | 1, 2 | 160 | |||
GenBank mRNA | DA657873.1 | - | 1, 2 | 160 |