Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1DP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027062.1 | 4 | 496 | |||
GenBank mRNA | DA657873.1 | - | 4 | 496 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY046836.1 | - | 3 | 416 | Chr. 13 - 31846721 - 31959579 [+] on Build GRCh38 |