Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1DP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027062.1 | 2 | 252 | |||
GenBank mRNA | BC021729.1 | - | 3 | 259 | ||
DA657873.1 | - | 2 | 252 |