Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027068.1 | 5 | 637 | |||
GenBank mRNA | BC105284.1 | - | 5 | 612 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU567964.1 | - | 5 | 611 | Chr. 7 - 105565126 - 105581500 [-] on Build GRCh38 |