Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024018.1 | 5 | 652 | |||
XM_017027427.1 | XP_016882916.1 | 5 | 667 | |||
XM_017027428.1 | XP_016882917.1 | 5 | 671 | |||
XM_017027429.1 | XP_016882918.1 | 5 | 698 | |||
GenBank mRNA | AK290435.1 | - | 5 | 507 | ||
BX640666.1 | - | 5 | 575 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA475980.1 | - | 5 | 507 | Chr. 19 - 34927048 - 34942663 [+] on Build GRCh38 |