Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015442.1 | 3 | 565 | |||
NR_024412.1 | 3 | 653 | ||||
GenBank mRNA | BC065765.1 | - | 3 | 541 | ||
BC107860.1 | - | 3 | 569 | |||
BX537645.1 | - | 3 | 538 |