Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026861.1 | 2 | 837 | |||
GenBank mRNA | CB270066.1 | - | 2 | 366 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA111262.1 | - | 2 | 482 | Chr. 6 - 165924256 - 165987684 [-] on Build GRCh38 |