Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026860.1 | 1 | 103 | |||
NR_026861.1 | 1 | 103 | ||||
GenBank mRNA | BC008632.1 | - | 1 | 103 | ||
BC039614.1 | - | 1 | 103 | |||
BX647106.1 | - | 1 | 112 | |||
DA416538.1 | - | 1 | 61 |
Gene Symbol: PDE10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535387.2 | XP_011533689.2 | 1 | 185 | ||
XM_017010194.1 | XP_016865683.1 | 1 | 185 | |||
XM_017010197.1 | XP_016865686.1 | 1 | 185 |