Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003108.1 | 1 | 75 | |||
GenBank mRNA | AL598622.1 | - | 1 | 98 | ||
CR936675.1 | - | 1 | 67 |