Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003108.1 | 2 | 211 | |||
GenBank mRNA | AL598622.1 | - | 2 | 234 | ||
CR936675.1 | - | 2 | 203 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF454870.1 | - | 2 | 181 | Chr. 17 - 43125614 - 43140040 [+] on Build GRCh38 |