Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_023915.1 | 1 | 825 | |||
GenBank mRNA | U12897.1 | - | LIT1674 IPW |
1 | 825 |