Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023915.1 | 3 | 4115 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR626616.1 | - | 1 | 1746 | Chr. 15 - 25120348 - 25122441 [+] on Build GRCh38 |