Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_023915.1 | 3 | 1391 | |||
GenBank mRNA | CN332356.1 | - | 3 | 523 | ||
U12897.1 | - | LIT1674 IPW |
3 | 1391 |
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832758.1 | - | 33 | 4735 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR000777.1 | - | 4 | 601 | Chr. 15 - 25115590 - 25119448 [+] on Build GRCh38 |